Learn More
Pipeline Packaging Corp 5G HDPE RECTANGULAR TH PAIL
SDP
Catalog No. NC3377356
Encompass_Preferred
Pipeline 5 GAL_NATURAL_HDPE_RECTANGULAR TH PAIL (RST)_3H1/Y1.8/100_@48/64_70 MM TE NECK,6 TPI,W/DUST CAP_NO VENT STEM, INTEG HANDLE (ALSO KNOWN AS JERRICANS).Purchase in multiples of 48 or 64.
Catalog No. NC3377356 Supplier Pipeline Packaging Corp Supplier No. 81248
Pipeline 5 GAL_NATURAL_HDPE_RECTANGULAR TH PAIL (RST)_3H1/Y1.8/100_@48/64_70 MM TE NECK,6 TPI,W/DUST CAP_NO VENT STEM, INTEG HANDLE (ALSO KNOWN AS JERRICANS).Purchase in multiples of 48 or 64.