Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, Isoquinoline-5-boronic acid, 96%, [371766-08-4], I2102 172.98 C9H8BNO2

Catalog No. 50982581
Encompass_Preferred

Compound: Isoquinoline-5-boronic acid

CAS#: 371766-08-4

MDL#: MFCD03839356

Background: Isoquinoline-5-boronic acid is used in Suzuki-Miyaura cross-coupling reactions pivotal to the formation of carbon-carbon bonds in complex organic molecule synthesis. It serves as a building block in pharmaceutical development targeting cancer and other diseases, and is used in bioconjugation, fluorescent probe development for biological imaging, and advanced materials research.

Catalog No. 50-982-581 Supplier Frontier Specialty Chemicals Supplier No. I21025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Isoquinoline-5-boronic acid

CAS#: 371766-08-4

MDL#: MFCD03839356

Background: Isoquinoline-5-boronic acid is used in Suzuki-Miyaura cross-coupling reactions pivotal to the formation of carbon-carbon bonds in complex organic molecule synthesis. It serves as a building block in pharmaceutical development targeting cancer and other diseases, and is used in bioconjugation, fluorescent probe development for biological imaging, and advanced materials research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.