Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, Methyl 3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl) propionate, 98%, [1150561-77-5], M10374 214.066 C10H19BO4

Catalog No. 50982660
Encompass_Preferred

Compound: Methyl 3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl) propionate

CAS#: 1150561-77-5

MDL#: MFCD10700159

Background: A methyl ester-functionalized pinacol boronate used as a Suzuki-Miyaura coupling partner in organic synthesis. The ester group provides a handle for later hydrolysis or amidation in pharmaceutical building block chemistry.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-982-660 Supplier Frontier Specialty Chemicals Supplier No. M103745G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Methyl 3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl) propionate

CAS#: 1150561-77-5

MDL#: MFCD10700159

Background: A methyl ester-functionalized pinacol boronate used as a Suzuki-Miyaura coupling partner in organic synthesis. The ester group provides a handle for later hydrolysis or amidation in pharmaceutical building block chemistry.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.