Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, n-Propylboronic acid, 98%, [17745-45-8], P6700 87.91 C3H9BO2

Catalog No. 50982982
Encompass_Preferred

Compound: n-Propylboronic acid

CAS#: 17745-45-8

MDL#: MFCD01074564

Background: n-Propylboronic acid is an alkylboronic acid used as a reagent in Suzuki cross-coupling reactions to form carbon-carbon bonds in pharmaceutical and agrochemical synthesis. It has been used to prepare mTOR inhibitors for anti-tumor research and endothelial lipase inhibitors. It has also been studied for its ability to inhibit serine proteases such as thrombin.

Catalog No. 50-982-982 Supplier Frontier Specialty Chemicals Supplier No. P67005G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: n-Propylboronic acid

CAS#: 17745-45-8

MDL#: MFCD01074564

Background: n-Propylboronic acid is an alkylboronic acid used as a reagent in Suzuki cross-coupling reactions to form carbon-carbon bonds in pharmaceutical and agrochemical synthesis. It has been used to prepare mTOR inhibitors for anti-tumor research and endothelial lipase inhibitors. It has also been studied for its ability to inhibit serine proteases such as thrombin.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.