Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, Naphthalene-2-boronic acid, 98%, [32316-92-0], N6183 171.99 C10H9BO2

Catalog No. 50982835
Encompass_Preferred

Compound: Naphthalene-2-boronic acid

CAS#: 32316-92-0

MDL#: MFCD00236051

Background: Naphthalene-2-boronic acid is a synthetic fine chemical used in the synthesis of pharmaceuticals and fine organic chemicals. It serves as a coupling partner in Suzuki-Miyaura and other organoboron cross-coupling reactions to form carbon-carbon bonds. It has also been used in the development of N-heterocyclic carbene ligands and organic light-emitting diode materials.

Storage: Store at 2-8°C under dry conditions.

Catalog No. 50-982-835 Supplier Frontier Specialty Chemicals Supplier No. N61835G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Naphthalene-2-boronic acid

CAS#: 32316-92-0

MDL#: MFCD00236051

Background: Naphthalene-2-boronic acid is a synthetic fine chemical used in the synthesis of pharmaceuticals and fine organic chemicals. It serves as a coupling partner in Suzuki-Miyaura and other organoboron cross-coupling reactions to form carbon-carbon bonds. It has also been used in the development of N-heterocyclic carbene ligands and organic light-emitting diode materials.

Storage: Store at 2-8°C under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.