Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, Phenylacetic acid-3-boronic acid pinacol ester, 98%, [797755-05-6], P1229 262.11 C14H19BO4

Catalog No. 50982913
Encompass_Preferred

Compound: Phenylacetic acid-3-boronic acid pinacol ester

CAS#: 797755-05-6

MDL#: MFCD04973389

Background: Phenylacetic acid-3-boronic acid pinacol ester, an isomer of the 4-substituted analog, is used as an organoboron building block in Suzuki coupling reactions for pharmaceutical synthesis. It provides a stable pinacol-protected boronic acid handle useful for constructing complex molecules bearing a carboxylic acid side chain.

Catalog No. 50-982-913 Supplier Frontier Specialty Chemicals Supplier No. P12295G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Phenylacetic acid-3-boronic acid pinacol ester

CAS#: 797755-05-6

MDL#: MFCD04973389

Background: Phenylacetic acid-3-boronic acid pinacol ester, an isomer of the 4-substituted analog, is used as an organoboron building block in Suzuki coupling reactions for pharmaceutical synthesis. It provides a stable pinacol-protected boronic acid handle useful for constructing complex molecules bearing a carboxylic acid side chain.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.