Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, Potassium vinyltrifluoroborate, 95%, [13682-77-4], P10254 133.95 C2H3BF3K

Catalog No. 50982860
Encompass_Preferred

Compound: Potassium vinyltrifluoroborate

CAS#: 13682-77-4

MDL#: MFCD02093335

Background: A stable, air- and moisture-tolerant vinyl organotrifluoroborate used as a coupling partner in Suzuki-type cross-coupling reactions. It is a versatile reagent for introducing vinyl groups into organic and pharmaceutical building blocks.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-982-860 Supplier Frontier Specialty Chemicals Supplier No. P102545G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Potassium vinyltrifluoroborate

CAS#: 13682-77-4

MDL#: MFCD02093335

Background: A stable, air- and moisture-tolerant vinyl organotrifluoroborate used as a coupling partner in Suzuki-type cross-coupling reactions. It is a versatile reagent for introducing vinyl groups into organic and pharmaceutical building blocks.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.