Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, Quinoline-3-boronic acid, 95%, [191162-39-7], Q5061 172.98 C9H8BNO2

Catalog No. 50983042
Encompass_Preferred

Compound: Quinoline-3-boronic acid

CAS#: 191162-39-7

MDL#: MFCD02183527

Background: Quinoline-3-boronic acid is a heteroaryl boronic acid used in Suzuki-Miyaura cross-coupling reactions to form carbon-carbon bonds. It has served as a reactant in the preparation of HIV protease inhibitors and has shown activity as an inhibitor of the Staphylococcus aureus NorA efflux pump. It is also used as a fluorescent sugar-sensing probe and as an intermediate in OLED materials.

Catalog No. 50-983-042 Supplier Frontier Specialty Chemicals Supplier No. Q50615G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Quinoline-3-boronic acid

CAS#: 191162-39-7

MDL#: MFCD02183527

Background: Quinoline-3-boronic acid is a heteroaryl boronic acid used in Suzuki-Miyaura cross-coupling reactions to form carbon-carbon bonds. It has served as a reactant in the preparation of HIV protease inhibitors and has shown activity as an inhibitor of the Staphylococcus aureus NorA efflux pump. It is also used as a fluorescent sugar-sensing probe and as an intermediate in OLED materials.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.