Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, Quinoline-5-boronic acid, 98%, [355386-94-6], Q1089 172.98 C9H8BNO2

Catalog No. 50983037
Encompass_Preferred

Compound: Quinoline-5-boronic acid

CAS#: 355386-94-6

MDL#: MFCD03095058

Background: Quinoline-5-boronic acid is used in the fabrication of organic semiconductors, where 5-substituted quinoline derivatives prepared via Suzuki coupling serve as electron-transport and hole-blocking materials in multilayer OLED architectures. It is also applied in the design of chemical sensors, in Suzuki-Miyaura coupling for drug discovery, and as a nitrogen donor ligand in metal-organic framework and fluorescent pH probe construction.

Catalog No. 50-983-037 Supplier Frontier Specialty Chemicals Supplier No. Q10895G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Quinoline-5-boronic acid

CAS#: 355386-94-6

MDL#: MFCD03095058

Background: Quinoline-5-boronic acid is used in the fabrication of organic semiconductors, where 5-substituted quinoline derivatives prepared via Suzuki coupling serve as electron-transport and hole-blocking materials in multilayer OLED architectures. It is also applied in the design of chemical sensors, in Suzuki-Miyaura coupling for drug discovery, and as a nitrogen donor ligand in metal-organic framework and fluorescent pH probe construction.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.