Learn More
US Plastic Corporation 5GAL FORT-PAK 10X10X15.25INEA
SDP
Catalog No. NC9009729
Encompass
This rectangular seamless container is made of tough, impact resistant, high density polyethylene resin. it is lightweight with a handle for easy pouring. Meets FDA standards. Not for use with fuels or oils. Not UN approved.
Catalog No. NC9009729 Supplier US Plastic Corporation Supplier No. 75023
This rectangular seamless container is made of tough, impact resistant, high density polyethylene resin. it is lightweight with a handle for easy pouring. Meets FDA standards. Not for use with fuels or oils. Not UN approved.