Learn More
Grainger Barrel Accessory and Supply Open-Head Pail, Polyethylene, 5 gal., White
Catalog No. NC1452239
Encompass_Preferred
This round 5 gal. open-head pail is constructed from durable plastic for reliable material handling. It is designed to be FDA compliant for versatile use and features a sturdy construction with a material thickness of 90 mil.
  • Volume capacity of 5 gal.
  • Maximum operating temperature of 150°F
  • FDA compliant material
  • Round shape with 11-1/4 in. diameter and 16 in. height
  • UN rating for solids: 1H2/X11.5/S
  • Not rated for liquids
Catalog No. NC1452239 Supplier Grainger Supplier No. 49EN59
This round 5 gal. open-head pail is constructed from durable plastic for reliable material handling. It is designed to be FDA compliant for versatile use and features a sturdy construction with a material thickness of 90 mil.
  • Volume capacity of 5 gal.
  • Maximum operating temperature of 150°F
  • FDA compliant material
  • Round shape with 11-1/4 in. diameter and 16 in. height
  • UN rating for solids: 1H2/X11.5/S
  • Not rated for liquids