Learn More
US Plastic Corporation 5GAL LIGHTWEIGHT CARBOY
SDP
Catalog No. NC0072913
Encompass
The Fort-Pak is a rigid plastic drum molded of high density polyethylene. Features a 2 in. closure and a center 3/4 in. NPT reducer in the cap. This translucent carboy meets FDA standards. Not for fuels or oils and has a 63mm neck.
Catalog No. NC0072913 Supplier US Plastic Corporation Supplier No. 75004
The Fort-Pak is a rigid plastic drum molded of high density polyethylene. Features a 2 in. closure and a center 3/4 in. NPT reducer in the cap. This translucent carboy meets FDA standards. Not for fuels or oils and has a 63mm neck.