Learn More
Cary Company 5 Gallon (20L) Natural Plastic Tight Head Container, 70mm (6TPI), Vent, UN Rated
SDP
Supplier:  Cary Company 56WTN5EA
Encompass
*THIS PART# IS ONLY FOR 1EA-47 EA. Quantities above 48 should be in even increments of 48 only. See mfg# 56WTN5 UN Rated. This 5 gallon (20 liter) natural high density polyethylene (HDPE) plastic tight head rectangular container features a 70mm (6TPI) neck finish and back vent cap. This pail comes with a fixed handle. Plastic closed head containers are ideal for storing liquids, paints, adhesives, additives, chemicals, and more.
Catalog No. NC3232621