Learn More
Cary Company 5 Gallon (20L) Natural Plastic Tight Head Container, 70mm (6TPI), Vent, UN Rated
SDP
Catalog No. NC3232418
Encompass
UN Rated, 100 PER CASE. This 5 gallon (20 liter) natural high density polyethylene (HDPE) plastic tight head rectangular container features a 70mm (6TPI) neck finish and back vent cap. This pail comes with a fixed handle. Plastic closed head containers are ideal for storing liquids, paints, adhesives, additives, chemicals, and more.
Catalog No. NC3232418 Supplier Cary Company Supplier No. 56WTN5
UN Rated, 100 PER CASE. This 5 gallon (20 liter) natural high density polyethylene (HDPE) plastic tight head rectangular container features a 70mm (6TPI) neck finish and back vent cap. This pail comes with a fixed handle. Plastic closed head containers are ideal for storing liquids, paints, adhesives, additives, chemicals, and more.