Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5H-1-Benzazepin-5-one, 1-(4-amino-2-methylbenzoyl)-7-chloro-1,2,3,4-tetrahydro- | 137977-97-0 | 99.9% | 328.80 | 500 MG
SDP

Catalog No. 5000403453
Encompass_Preferred

Tolvaptan Impurity from MedChemExpress is a chemical compound with the molecular formula C18H17ClN2O2 and a molecular weight of 328.80. It is provided as a white to off-white solid with a purity of 99.92%. This product is intended for research use only and has not been fully validated for medical applications.

  • Provided as a white to off-white solid
  • Purity of 99.92%
  • Suitable for research applications
  • Store as powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-03453 Supplier Medchemexpress LLC Supplier No. HY42476500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tolvaptan Impurity from MedChemExpress is a chemical compound with the molecular formula C18H17ClN2O2 and a molecular weight of 328.80. It is provided as a white to off-white solid with a purity of 99.92%. This product is intended for research use only and has not been fully validated for medical applications.

  • Provided as a white to off-white solid
  • Purity of 99.92%
  • Suitable for research applications
  • Store as powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.