Learn More
Medchemexpress LLC 5H-benzo[g]-1,3-benzodioxolo[6,5,4-de]quinoline, 6,7,7a,8-tetrahydro-7-methyl (7aR) | 548-08-3 | 99.9% | 10 MG
SDP
Catalog No. 5000232094
Encompass_Preferred
Roemerine is an aporphine alkaloid small-molecule reagent used in biochemical and pharmacological research. It is supplied in small-mass packs for in vitro studies and is commonly used to probe P-glycoprotein interactions and serotonin receptor activity.
  • High purity suitable for analytical and biological assays.
  • Available in small pack sizes, including 10 MG.
  • Molecular weight 279.33 g/mol and formula C18H17NO2 for reference.
  • Store sealed at 4°C; follow low-temperature storage guidelines for solutions.
Catalog No. 50-002-32094 Supplier Medchemexpress LLC Supplier No. HY12179310MG
Roemerine is an aporphine alkaloid small-molecule reagent used in biochemical and pharmacological research. It is supplied in small-mass packs for in vitro studies and is commonly used to probe P-glycoprotein interactions and serotonin receptor activity.
  • High purity suitable for analytical and biological assays.
  • Available in small pack sizes, including 10 MG.
  • Molecular weight 279.33 g/mol and formula C18H17NO2 for reference.
  • Store sealed at 4°C; follow low-temperature storage guidelines for solutions.