Learn More
Medchemexpress LLC 5H-benzo[g]-1,3-benzodioxolo[6,5,4-de]quinoline, 6,7,7a,8-tetrahydro-7-methyl- (7aR) | 548-08-3 | 99.9% | 5 MG
SDP
Catalog No. 5000230584
Encompass_Preferred
Roemerine is an isoquinoline alkaloid supplied as a white to off-white solid for research use. It has molecular formula C18H17NO2, a molecular weight of 279.33 g/mol, and analytical purity of 99.89% (HPLC). The compound is soluble in DMSO (100 mg/mL) and should be stored sealed, away from moisture and light.
  • Isoquinoline alkaloid with reported antibacterial and CNS activity.
  • High purity (99.9% by HPLC) suitable for analytical applications.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic.
  • Chemical identity confirmed by 1H NMR and MS.
  • Store sealed at 4°C; in solvent, -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-002-30584 Supplier Medchemexpress LLC Supplier No. HY1217935MG
Roemerine is an isoquinoline alkaloid supplied as a white to off-white solid for research use. It has molecular formula C18H17NO2, a molecular weight of 279.33 g/mol, and analytical purity of 99.89% (HPLC). The compound is soluble in DMSO (100 mg/mL) and should be stored sealed, away from moisture and light.
  • Isoquinoline alkaloid with reported antibacterial and CNS activity.
  • High purity (99.9% by HPLC) suitable for analytical applications.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic.
  • Chemical identity confirmed by 1H NMR and MS.
  • Store sealed at 4°C; in solvent, -80°C for 6 months or -20°C for 1 month.