Learn More
Medchemexpress LLC 5H-Dibenz[b,f]azepine, 10,11-dihydro- | 494-19-9 | 99.98% | 25 G
SDP
Catalog No. 5000413914
Encompass_Preferred
Iminodibenzyl is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It serves as an intermediate for the antiepileptic agent Carbamazepine and has been shown to inhibit the 5-lipoxygenase pathway in rat basophilic leukemia (RBL-1) cells. This product is intended for research use only.
  • Suitable for life science research
  • Used as an intermediate for antiepileptic agent Carbamazepine
  • Inhibits 5-lipoxygenase pathway in RBL-1 cells
Catalog No. 50-004-13914 Supplier Medchemexpress LLC Supplier No. 25GHYB2131
Iminodibenzyl is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It serves as an intermediate for the antiepileptic agent Carbamazepine and has been shown to inhibit the 5-lipoxygenase pathway in rat basophilic leukemia (RBL-1) cells. This product is intended for research use only.
  • Suitable for life science research
  • Used as an intermediate for antiepileptic agent Carbamazepine
  • Inhibits 5-lipoxygenase pathway in RBL-1 cells