Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5H-dibenzo[b,e][1,4]diazepine, 8-chloro-11-(4-ethyl-1-piperazinyl)-1-fluoro- | 2369979-67-7 | MFCD34167518 | 99.5% | C19H20ClFN4 | 10MG
SDP

Catalog No. 5000261695
Encompass_Preferred

JHU37152 is a potent, brain-penetrant DREADD agonist selective for hM3Dq and hM4Di receptors. In HEK-293 assays it exhibits EC50s of 5 nM (hM3Dq) and 0.5 nM (hM4Di), displaces [3H]clozapine, and is supplied as a high-purity research reagent.

  • High purity: 99.45%.
  • Cas number: 2369979-67-7.
  • Chemical formula: C19H20ClFN4.
  • Molecular weight: 358.84 g·mol⁻1.
  • Physical state: solid, white to yellow.
  • Solubility: soluble in DMSO (~33.33 mg/mL).
  • Storage: powder -20°C for long term; in solvent -80°C recommended.
  • Applications: in vitro and in vivo neuroscience research.

Catalog No. 50-002-61695 Supplier Medchemexpress LLC Supplier No. HY13189110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JHU37152 is a potent, brain-penetrant DREADD agonist selective for hM3Dq and hM4Di receptors. In HEK-293 assays it exhibits EC50s of 5 nM (hM3Dq) and 0.5 nM (hM4Di), displaces [3H]clozapine, and is supplied as a high-purity research reagent.

  • High purity: 99.45%.
  • Cas number: 2369979-67-7.
  • Chemical formula: C19H20ClFN4.
  • Molecular weight: 358.84 g·mol⁻1.
  • Physical state: solid, white to yellow.
  • Solubility: soluble in DMSO (~33.33 mg/mL).
  • Storage: powder -20°C for long term; in solvent -80°C recommended.
  • Applications: in vitro and in vivo neuroscience research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.