Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5H-imidazo[2,1-b]oxazine, 6,7-dihydro-2-nitro-6-[[6-[4-(trifluoromethoxy)phenyl]-3-pyridinyl]methoxy] | 1257426-19-9 | 99.7% | 436.34 | C19H15F3N4O5 | 100MG
SDP

Catalog No. 5000236436
Encompass_Preferred

TBA-354 is a nitroimidazole-class anti-tuberculosis research compound with reported activity against Mycobacterium tuberculosis, including some monoresistant strains. It is provided as a high-purity solid and as ready-to-use DMSO solutions for use in antimicrobial screening and efficacy studies.

  • Molecular formula C19H15F3N4O5.
  • Molecular weight 436.34.
  • CAS number 1257426-19-9.
  • Purity 99.7%.
  • Supplied as solid and as 10 mM solution in DMSO.
  • Available in small-scale quantities from 5 mg up to 500 mg.
  • Demonstrates bactericidal activity against M. tuberculosis strains.
  • Intended for research use in in vitro and in vivo studies.

Catalog No. 50-002-36436 Supplier Medchemexpress LLC Supplier No. HY12485100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TBA-354 is a nitroimidazole-class anti-tuberculosis research compound with reported activity against Mycobacterium tuberculosis, including some monoresistant strains. It is provided as a high-purity solid and as ready-to-use DMSO solutions for use in antimicrobial screening and efficacy studies.

  • Molecular formula C19H15F3N4O5.
  • Molecular weight 436.34.
  • CAS number 1257426-19-9.
  • Purity 99.7%.
  • Supplied as solid and as 10 mM solution in DMSO.
  • Available in small-scale quantities from 5 mg up to 500 mg.
  • Demonstrates bactericidal activity against M. tuberculosis strains.
  • Intended for research use in in vitro and in vivo studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.