Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5H-Perfluoropentanoic acid, 96%

Catalog No. AAL1696014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1696014 25 g
AAL1696006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1696014 Supplier Thermo Scientific Chemicals Supplier No. L1696014

CAS: 376-72-7 | C5H2F8O2 | 246.056 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376-72-7
Molecular Formula C5H2F8O2
Molecular Weight (g/mol) 246.056
MDL Number MFCD00054690
InChI Key VGFKXVSMDOKOJZ-UHFFFAOYSA-N
Synonym 5h-octafluoropentanoic acid, octafluoropentanoic acid, 2,2,3,3,4,4,5,5-octafluorovaleric acid, 5h-perfluoropentanoic acid, omega-h-oktafluorpentansaeure german, pentanoic acid, 2,2,3,3,4,4,5,5-octafluoro, omega-h-oktafluorpentansaeure, 5-h-octafluoropentanoic acid, 4-02-00-00877 beilstein handbook reference
PubChem CID 120227
IUPAC Name 2,2,3,3,4,4,5,5-octafluoropentanoic acid
SMILES C(C(C(C(C(=O)O)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 0°C
Density 1.725
Boiling Point 165°C to 166°C
Refractive Index 1.319
Quantity 25 g
UN Number UN3265
Beilstein 1797233
Formula Weight 246.06
Percent Purity 96%
Chemical Name or Material 5H-Perfluoropentanoic acid
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 206-812-3
RTECSNumber YV7799000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.