Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5H-pyrano[3,2-d]oxazole-6,7-diol diacetate (stereoisomer) | 35954-65-5 | 99.5% | 329.30 g/mol | C14H19NO8 | 100MG
SDP

Catalog No. 5000239270
Encompass_Preferred

FR054 is a small-molecule inhibitor of the hexosamine biosynthetic pathway enzyme PGM3, studied for anti-breast cancer activity in preclinical research. It is supplied as a purified oil for research use only and is accompanied by analytical documentation supporting identity and purity.

  • Inhibits PGM3, relevant to hexosamine biosynthetic pathway studies.
  • High purity (99.5%) suitable for preclinical experiments.
  • Molecular formula C14H19NO8; molecular weight 329.30 g/mol.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Storage: sealed at -20°C; in solution store at -80°C up to 6 months.
  • Provided as an oil suitable for in vitro assays.
  • Analytical data available: COA, H NMR, MS, RP-HPLC.

Catalog No. 50-002-39270 Supplier Medchemexpress LLC Supplier No. HY124909A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FR054 is a small-molecule inhibitor of the hexosamine biosynthetic pathway enzyme PGM3, studied for anti-breast cancer activity in preclinical research. It is supplied as a purified oil for research use only and is accompanied by analytical documentation supporting identity and purity.

  • Inhibits PGM3, relevant to hexosamine biosynthetic pathway studies.
  • High purity (99.5%) suitable for preclinical experiments.
  • Molecular formula C14H19NO8; molecular weight 329.30 g/mol.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Storage: sealed at -20°C; in solution store at -80°C up to 6 months.
  • Provided as an oil suitable for in vitro assays.
  • Analytical data available: COA, H NMR, MS, RP-HPLC.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.