Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5H-Pyrido[4,3-b]indole 98.0+%, TCI America™
SDP

Catalog No. P2362200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2362200MG 200 mg
P23621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2362200MG Supplier TCI America Supplier No. P2362200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 244-69-9
Molecular Formula C11H8N2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00965976
InChI Key RDMFHRSPDKWERA-UHFFFAOYSA-N
Synonym gamma-Carboline
PubChem CID 130802
IUPAC Name 5H-pyrido[4,3-b]indole
SMILES N1C2=C(C=CC=C2)C2=C1C=CN=C2

Specifications

Melting Point 233°C
Quantity 200 mg
Formula Weight 168.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5H-Pyrido[4,3-b]indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.