Learn More
Cayman Chemical 5hydroxy SaxaglIptInhydroc
SDP
Catalog No. 502936285
Encompass_Preferred
A major active metabolite of saxagliptin; inhibits DPP-4 (Ki = 2.6 nM for human recombinant enzyme); selective for DPP-4 over DPP-8 and DPP-9 (Kis = 2,495 and 423 nM, respectively, for human recombinant enzymes)
Catalog No. 50-293-6285 Supplier Cayman Chemical Supplier No. 26710500 UG
A major active metabolite of saxagliptin; inhibits DPP-4 (Ki = 2.6 nM for human recombinant enzyme); selective for DPP-4 over DPP-8 and DPP-9 (Kis = 2,495 and 423 nM, respectively, for human recombinant enzymes)