Learn More
Cayman Chemical 5-Hydroxymethyl-2-furancarboxylic Acid, Purity: ≥98%, CAS: 6338-41-6, 1 g
SDP
Catalog No. 502994998
Encompass_Preferred
A major metabolite of 5-hydroxymethyl-2-furfural; can be used as a building block in the enzymatic synthesis of macrocyclic oligoesters
Catalog No. 50-299-4998 Supplier Cayman Chemical Supplier No. 229991 G
A major metabolite of 5-hydroxymethyl-2-furfural; can be used as a building block in the enzymatic synthesis of macrocyclic oligoesters