Learn More
Biomatik Corporation 5-Hydroxytryptamine (5-HT) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12286
Encompass_Preferred
5-Hydroxytryptamine (5-HT) Mini sample ELISA Kit for detecting 5-HT in serum, plasma. Species Reactivity: Universal. Detection Range: 11.11-900ng/ml. Sensitivity: 3.93ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: 5HT; Serotonin; N-Methyl-Gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6369