Learn More
Sklar Instruments Forceps, Econo Thumb Tissue Forceps, Sklar 5 in., 1x2 Teeth, Straight, Thumb, Stainless Steel, BX/12.
SDP
Catalog No. 501098101 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Econo Thumb Tissue Forceps, Sklar, 5 in., 1x2 Teeth, Straight, Thumb, Stainless Steel, BX/12. Econo Thumb Tissue Forceps are used for grasping, holding, and manipulating various types of tissues in multiple procedures. These instruments have spring tension shanks that ultimately determine how far the instrument will open and the thickness of the tissue that can be held. Some thumb tissue forceps have lesser tension and can only hold fine membranes or single layer tissue. Others have greater tension and can hold thick, heavy tissue. Thumb tissue forceps are commonly toothed in order to grip the tissue more firmly than serrated tips alone. They are available in several lengths to accommodate specific procedures, tissue types, and user preference. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with 1x2 teeth and a length of 5 inches. They are sold in boxes of 12.
Catalog No. 50-109-8101 Supplier Sklar Instruments Supplier No. 21757
Forceps, Econo Thumb Tissue Forceps, Sklar, 5 in., 1x2 Teeth, Straight, Thumb, Stainless Steel, BX/12. Econo Thumb Tissue Forceps are used for grasping, holding, and manipulating various types of tissues in multiple procedures. These instruments have spring tension shanks that ultimately determine how far the instrument will open and the thickness of the tissue that can be held. Some thumb tissue forceps have lesser tension and can only hold fine membranes or single layer tissue. Others have greater tension and can hold thick, heavy tissue. Thumb tissue forceps are commonly toothed in order to grip the tissue more firmly than serrated tips alone. They are available in several lengths to accommodate specific procedures, tissue types, and user preference. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with 1x2 teeth and a length of 5 inches. They are sold in boxes of 12.