Learn More
Sklar Instruments Forceps, Semken Tissue Forceps, Sklar 5 in., Straight, Serrated, Thumb, Stainless Steel, EA.
SDP
Catalog No. 501098144 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Semken Tissue Forceps, Sklar, 5 in., Straight, Serrated, Thumb, Stainless Steel, EA. Sklar Semken Tissue Forceps are lightweight, narrow, toothed forceps used for handling primarily fine membranes or single layer tissues. This product is straight with a smooth, round tip, and overall length of 5 inches.
Catalog No. 50-109-8144 Supplier Sklar Instruments Supplier No. 224157
Forceps, Semken Tissue Forceps, Sklar, 5 in., Straight, Serrated, Thumb, Stainless Steel, EA. Sklar Semken Tissue Forceps are lightweight, narrow, toothed forceps used for handling primarily fine membranes or single layer tissues. This product is straight with a smooth, round tip, and overall length of 5 inches.