Learn More
Chemglass Life Sciences Chemglass 5 L Two-Neck Receiving Flask, 45/50 Ground-Glass Joints (for 10 L Reactor System)
SDP
Catalog No. 502733558
Encompass_Preferred
The Chemglass 5 L two-neck receiving flask is a glass vessel designed to collect distillate or reaction products in reactor and distillation setups. It features two 45/50 ground-glass joints and is specified as the receiving flask for 10 L reactor/distillation systems.
  • 5 L capacity for collecting distillate and reaction products.
  • Two necks to accommodate condensers, adapters, or sensors.
  • 45/50 ground-glass joints for secure, standard connections.
  • Designed for use with 10 L reactor and distillation systems.
  • Suitable for standard laboratory distillation and reactor receiving applications.
Catalog No. 50-273-3558 Supplier Chemglass Life Sciences Supplier No. CG1965D220
The Chemglass 5 L two-neck receiving flask is a glass vessel designed to collect distillate or reaction products in reactor and distillation setups. It features two 45/50 ground-glass joints and is specified as the receiving flask for 10 L reactor/distillation systems.
  • 5 L capacity for collecting distillate and reaction products.
  • Two necks to accommodate condensers, adapters, or sensors.
  • 45/50 ground-glass joints for secure, standard connections.
  • Designed for use with 10 L reactor and distillation systems.
  • Suitable for standard laboratory distillation and reactor receiving applications.