Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 5L, 34/45 center, 24/40 sides
SDP
Catalog No. 509557536
Encompass_Preferred
5L round bottom flask with four necks, arranged three in-line and one at 90. Center neck has a 34/45 joint, and angled side necks have 24/40 joints.
Catalog No. 50-955-7536 Supplier Ace Glass, Inc. Supplier No. 695256
5L round bottom flask with four necks, arranged three in-line and one at 90. Center neck has a 34/45 joint, and angled side necks have 24/40 joints.