Learn More
Chemglass Life Sciences Flask, Round Bottom, 5000mL, Heavy Wall, 5-Neck, 24/40 CN - 24/40 SN's, Vertical
SDP
Catalog No. CG153506
Encompass_Preferred
Flask, Round Bottom, 5000mL, Heavy Wall, 24/40 - 24/40, 5-Neck, Vertical. Round bottom, HEAVY WALL, five-neck flask with standard taper outer joints on the center neck and vertical side necks.
Catalog No. CG153506 Supplier Chemglass Life Sciences Supplier No. CG153506
Flask, Round Bottom, 5000mL, Heavy Wall, 24/40 - 24/40, 5-Neck, Vertical. Round bottom, HEAVY WALL, five-neck flask with standard taper outer joints on the center neck and vertical side necks.