Learn More
Foxx Life Sciences EZgrip 5L (1 Gal) Light Blocking Amber HDPE Space Saving Carboy with Leakproof Spigot, 83mm (83B) VersaCap and 6.9L Max Capacity, Large Pinched Handles
Catalog No. 501178741
Encompass_Preferred
15K-1211-OEM: Carboy, 5L, Amber HDPE, 83B Cap, w Spigot. Foxx Life Sciences EZgrip Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the container and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric graduation marks which are certified to 5% accuracy. Recessed spigot location allows for Carboys to be stored on any surface without requiring overhang, and the Foxx Life Sciences Spigot is 100% leakproof. 100% Leak-Proof Spigot available in certain models. EZfil wide-mouth design for easy filling and cleaning. VersaCap offers a wide variety of top connection options.
Catalog No. 50-117-8741 Supplier Foxx Life Sciences Supplier No. 15K1211OEM
15K-1211-OEM: Carboy, 5L, Amber HDPE, 83B Cap, w Spigot. Foxx Life Sciences EZgrip Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the container and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric graduation marks which are certified to 5% accuracy. Recessed spigot location allows for Carboys to be stored on any surface without requiring overhang, and the Foxx Life Sciences Spigot is 100% leakproof. 100% Leak-Proof Spigot available in certain models. EZfil wide-mouth design for easy filling and cleaning. VersaCap offers a wide variety of top connection options.