Learn More
Bellco Glass, Inc. 5L BCS Flat Bottom System,115V
SDP
Catalog No. 501946253
Encompass_Preferred
Bellco Control System, Flat Bottom. The Bellco Stainless Steel Shaft ConversionKit is designed to convert your standard glass shaft to stainless steel on 1967 and 1965 Spinner Flasks. Sizes range from:--100 mL to 36 liters for 1965 spinners--100 to 3000 mL for 1967 spinner flasksMaterial: --316L Stainless Steel 1965 & 1967 Series Micro-Carrier Spinner Flask 5L BCS Flat Bottom System 115v. MADE IN THE USA
Catalog No. 50-194-6253 Supplier Bellco Glass, Inc. Supplier No. 780005510
Add to Cart
Add to Cart
Request a Quote
Bellco Control System, Flat Bottom. The Bellco Stainless Steel Shaft ConversionKit is designed to convert your standard glass shaft to stainless steel on 1967 and 1965 Spinner Flasks. Sizes range from:--100 mL to 36 liters for 1965 spinners--100 to 3000 mL for 1967 spinner flasksMaterial: --316L Stainless Steel 1965 & 1967 Series Micro-Carrier Spinner Flask 5L BCS Flat Bottom System 115v. MADE IN THE USA