Learn More
Chemglass Life Sciences Chemglass 5L Bio Reactor Baffle Assembly, M10 (for 5 L Bioreactors)
SDP
Catalog No. 502733316
Encompass
The Chemglass 5L Bio Reactor Baffle Assembly, M10, is a stainless steel replacement baffle designed to improve mixing efficiency in 5 L glass bioreactors. It mounts near the reactor wall to reduce vortexing and enhance gas-liquid contact for microbial and cell culture applications. This assembly is offered as a like-for-like replacement for common bioreactor systems.
  • Stainless steel 316L construction with electropolished finish for corrosion resistance.
  • M10 threaded mount sized for 5 L reactor vessels.
  • Improves mixing and reduces vortex formation to enhance mass transfer.
  • Serves as a like-for-like replacement for common bioreactor systems.
Catalog No. 50-273-3316 Supplier Chemglass Life Sciences Supplier No. CLS138210
The Chemglass 5L Bio Reactor Baffle Assembly, M10, is a stainless steel replacement baffle designed to improve mixing efficiency in 5 L glass bioreactors. It mounts near the reactor wall to reduce vortexing and enhance gas-liquid contact for microbial and cell culture applications. This assembly is offered as a like-for-like replacement for common bioreactor systems.
  • Stainless steel 316L construction with electropolished finish for corrosion resistance.
  • M10 threaded mount sized for 5 L reactor vessels.
  • Improves mixing and reduces vortex formation to enhance mass transfer.
  • Serves as a like-for-like replacement for common bioreactor systems.