Learn More
Ace Glass, Inc. Lab bottle, Duran, 5L, safety coated, pouring ring & PTFE lined GL 45 PBT cap, cs/1
SDP
Catalog No. NC0697913
Encompass
Chemical-resistant and stable 5L Duran laboratory bottles, fabricated from borosilicate glass and safety coated. Supplied graduated with PTFE-lined GL45 PBT cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/1.
Catalog No. NC0697913 Supplier Ace Glass, Inc. Supplier No. 5539130
Chemical-resistant and stable 5L Duran laboratory bottles, fabricated from borosilicate glass and safety coated. Supplied graduated with PTFE-lined GL45 PBT cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/1.