Learn More
Chemglass Life Sciences Nest 5 L High-efficiency Polycarbonate Erlenmeyer Flask, Sterile, Vent Cap with 0.2 μm Filter
SDP
Catalog No. 502733455
Encompass
High-efficiency 5 L Erlenmeyer flask for scale-up suspension and shaker cultures. Made from high-clarity polycarbonate and supplied sterile, the flask provides gas exchange through an HDPE vent cap with a 0.2 μm hydrophobic filter and includes a clear molded graduation and sturdy handles for easy handling.
  • 5 L capacity suitable for scale-up suspension and shaker cultures.
  • High-clarity polycarbonate construction for visibility and durability.
  • Sterilized by electron beam; non-pyrogenic and ready to use.
  • HDPE vent cap with 0.2 μm hydrophobic filter for gas exchange.
  • Clear molded graduation scale and sturdy handles for handling.
  • Packaged sterile in bags, four flasks per case.
  • Recommended working volume: 30-40% of flask capacity.
  • Suggested rotation start speed: 75-125 RPM; adjust per application.
  • Bottle dimensions: 90 mm neck diameter; 285 mm height; 230 mm bottom diameter.
Catalog No. 50-273-3455 Supplier Chemglass Life Sciences Supplier No. CGN21655LSC
Add to Cart
Add to Cart
High-efficiency 5 L Erlenmeyer flask for scale-up suspension and shaker cultures. Made from high-clarity polycarbonate and supplied sterile, the flask provides gas exchange through an HDPE vent cap with a 0.2 μm hydrophobic filter and includes a clear molded graduation and sturdy handles for easy handling.
  • 5 L capacity suitable for scale-up suspension and shaker cultures.
  • High-clarity polycarbonate construction for visibility and durability.
  • Sterilized by electron beam; non-pyrogenic and ready to use.
  • HDPE vent cap with 0.2 μm hydrophobic filter for gas exchange.
  • Clear molded graduation scale and sturdy handles for handling.
  • Packaged sterile in bags, four flasks per case.
  • Recommended working volume: 30-40% of flask capacity.
  • Suggested rotation start speed: 75-125 RPM; adjust per application.
  • Bottle dimensions: 90 mm neck diameter; 285 mm height; 230 mm bottom diameter.