Learn More
Chemglass Life Sciences 5L Erlenmeyer Flask, High Efficiency, Vent Cap, Sterile, Nest
SDP
Catalog No. 501941157
Encompass
5L Erlenmeyer Flask, Vent Cap, Sterile, Nest. 5L Erlenmeyer flask is designed to help scale-up suspension culture with space efficiency in mind. The flasks are sterile and ready-to-use, ideal for shaker culture applications, and storage. Polycarbonate flask with HDPE vent cap with 0.2¿m hydrophobic filter for gas exchange without contamination. Bottle dimensions: 90mm Bottle Neck Diameter, 285mm H, 230mm Bottom Diameter. Polycarbonate has high standard of transparency and the flask has a clear graduation scale for easy observation. Sterilized by E-Beam. Non-Pyrogenic. Packaged in sterile bags, 4 per case.
Catalog No. 50-194-1157 Supplier Chemglass Life Sciences Supplier No. CGN21655L
5L Erlenmeyer Flask, Vent Cap, Sterile, Nest. 5L Erlenmeyer flask is designed to help scale-up suspension culture with space efficiency in mind. The flasks are sterile and ready-to-use, ideal for shaker culture applications, and storage. Polycarbonate flask with HDPE vent cap with 0.2¿m hydrophobic filter for gas exchange without contamination. Bottle dimensions: 90mm Bottle Neck Diameter, 285mm H, 230mm Bottom Diameter. Polycarbonate has high standard of transparency and the flask has a clear graduation scale for easy observation. Sterilized by E-Beam. Non-Pyrogenic. Packaged in sterile bags, 4 per case.