Learn More
Foxx Life Sciences EZBio 5L Erlenmeyer Polycarbonate (PC) Flasks - 90mm Closed Cap w/Handle - Sterilized - 4/CS
GREENER_CHOICE
Catalog No. NC1818233
Encompass_Preferred
The EZBio 5L Erlenmeyer flask is designed to be space-efficient with scalability in mind. The flasks come E-Beam sterilized and ready-to-use for shaker culture applications, microbial cultures, flora growth, media preparation, storage, and all related sterile applications. They fit all standard shaking incubator clamps and can be placed on any platform. The High-Density Polyethylene (HDPE) closed cap and handles allow for sturdy transport and ease of use. 
Features
Vessels are constructed from polycarbonate (PC bottle).
High standard of transparency and clear scale, easy to observe.
Sturdy, good for transport.
Nonpyrogenic and DNase/RNase free
Sterilized by E-Beam
Note During liquid shaking, be sure to pay attention to the water level. During gas shaking, be sure to pay attention to the temperature. 
Catalog No. NC1818233 Supplier Foxx Life Sciences Supplier No. 568FD01FLS
The EZBio 5L Erlenmeyer flask is designed to be space-efficient with scalability in mind. The flasks come E-Beam sterilized and ready-to-use for shaker culture applications, microbial cultures, flora growth, media preparation, storage, and all related sterile applications. They fit all standard shaking incubator clamps and can be placed on any platform. The High-Density Polyethylene (HDPE) closed cap and handles allow for sturdy transport and ease of use. 
Features
Vessels are constructed from polycarbonate (PC bottle).
High standard of transparency and clear scale, easy to observe.
Sturdy, good for transport.
Nonpyrogenic and DNase/RNase free
Sterilized by E-Beam
Note During liquid shaking, be sure to pay attention to the water level. During gas shaking, be sure to pay attention to the temperature. 