Learn More
HCL Labels 16.9 oz (.5L) Pre-labeled GHS Compliant Dual-Action Kwazar Spray Bottle Ethanol 80%, Bottle Supplied Empty, 1 EA
SDP
Catalog No. 501633555
Encompass_Preferred
Bottles, HCL Labels, Inc., .5L, GHS Ethanol 80% Spray Bottle, These Kwazar bottles are pre-labeled with a GHS hazard communication chemical label. Bottles are chemical resistant and empty.
Catalog No. 50-163-3555 Supplier HCL Labels Supplier No. GHSKWBOT0009
Bottles, HCL Labels, Inc., .5L, GHS Ethanol 80% Spray Bottle, These Kwazar bottles are pre-labeled with a GHS hazard communication chemical label. Bottles are chemical resistant and empty.