Learn More
Bellco Glass, Inc. 5L Flat Bottom Omni Vessel Large Flange
SDP
Catalog No. 501944877
Encompass_Preferred
Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. MADE IN THE USA. 5l flat bottom 215 x 250mm
Catalog No. 50-194-4877 Supplier Bellco Glass, Inc. Supplier No. 196488050
Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. MADE IN THE USA. 5l flat bottom 215 x 250mm