Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC0834175
Encompass_Preferred
Hemispherical fabric heating mantle for 5L single or multi neck flasks to 400 degrees C maximum internal operating temperature, 600W, 115V. CSA
Catalog No. NC0834175 Supplier Glas Col LLC Supplier No. 100A O414
Hemispherical fabric heating mantle for 5L single or multi neck flasks to 400 degrees C maximum internal operating temperature, 600W, 115V. CSA