Learn More
Alkali Scientific 5L High Efficiency Erlenmeyer
SDP
Supplier:  Alkali Scientific EFC442
Encompass_Preferred
Bi-directional Transfer Cap for 2/3/5L high efficient Erlenmeyer flasks The liquid transfer is achieved by the connection of a peristaltic pump between the Erlenmeyer flask and the device Upon completion of the transfer the transfer cap can be replaced with a vent cap for culture The pipe for liquid transfer is outfitted with Male MPC Connector or the Luer Taper and the corresponding Seal Cap which makes it convenient for customers to connect the ends of pipes with the same connector Besides the tube at the bottom can reach the flask bottom then completely and aseptically transfer the liquid Meanwhile contamination risk is greatly reduced due to the simplified aseptic transfer procedures with the Bi-directional Transfer Cap for Erlenmeyer Flask Product Features Closed Liquid Transfer reduces the contamination risk during the operation procedures The vent cap with 0 2um membrane is permeable and waterproof thus can effectively block the microorganisms and avoids the contaminatio
Catalog No. 50-302-7561