Learn More
Ace Glass, Inc. Beaker, 5L, jacketed, glass hose connections for 7/16 or 1/2intubing, 250mm inside height, 160mm inside diameter
SDP
Catalog No. NC0421822
Encompass_Preferred
5L jacketed beaker with one upper and one lower glass hose connection on opposite sides, for 7/16 in. or 1/2 in. tubing. Dimensions: 250mm I.H. x 160mm I.D. Beaker does not have a pouring spout.
Catalog No. NC0421822 Supplier Ace Glass, Inc. Supplier No. 534035
Add to Cart
Add to Cart
5L jacketed beaker with one upper and one lower glass hose connection on opposite sides, for 7/16 in. or 1/2 in. tubing. Dimensions: 250mm I.H. x 160mm I.D. Beaker does not have a pouring spout.