Learn More
Structure Probe Inc Liquid Nitrogen Dewar, 5 liter, Neck Diameter 5.6 inch
SDP
Catalog No. NC9755323
Encompass
SPI Liquid Nitrogen Dewars, 5L, 17.5 in. H, 7.6 in. dia., Neck Diameter: 5.6 in. , Empty Weight: 5 lb., Full Weight: 14 lb., Evaporation Loss: 0.77L/day
Catalog No. NC9755323 Supplier Structure Probe Inc Supplier No. 10416AB
SPI Liquid Nitrogen Dewars, 5L, 17.5 in. H, 7.6 in. dia., Neck Diameter: 5.6 in. , Empty Weight: 5 lb., Full Weight: 14 lb., Evaporation Loss: 0.77L/day