Learn More
Chemglass Life Sciences 5L Media Transfer Vessel, Vent Cap, w/Barb and #24 C-Flex Tubing, Plugged, Sterile
SDP
Supplier:  Chemglass Life Sciences CLS11715LWB
Encompass
Dual purpose, 5L transfer flasks are designed for the sterile transfer of liquids as well as for single-use inoculation vessels.They’re ideal for use doing gravity feed from sterile, 5L vessels.Erlenmeyer-style, polycarbonate flask includes a cap with 0. 2 um hydrophobic filter, 1/4” barbed fitting, 1/4” ID x 3/8” OD or 1/4” ID x 7/16” OD weldable C-Flex  tubing with a plugged end and tubing clamp.Supplied gamma irradiated, sterile and individually double bagged.
Catalog No. NC2263135