Learn More
Chemglass Life Sciences 5L Media Transfer Vessel, Vent Cap, w/Barb and #17 C-Flex Tubing, Plugged, Sterile
SDP
Catalog No. NC2038281
Encompass
Dual purpose, 5L transfer flasks are designed for the sterile transfer of liquids as well as for single-use inoculation vessels.They’re ideal for use doing gravity feed from sterile, 5L vessels.Erlenmeyer-style, polycarbonate flask includes a cap with 0. 2 um hydrophobic filter, 1/4” barbed fitting, 1/4” ID x 3/8” OD or 1/4” ID x 7/16” OD weldable C-Flex  tubing with a plugged end and tubing clamp.Supplied gamma irradiated, sterile and individually double bagged.
Catalog No. NC2038281 Supplier Chemglass Life Sciences Supplier No. CLS11715LWT
Dual purpose, 5L transfer flasks are designed for the sterile transfer of liquids as well as for single-use inoculation vessels.They’re ideal for use doing gravity feed from sterile, 5L vessels.Erlenmeyer-style, polycarbonate flask includes a cap with 0. 2 um hydrophobic filter, 1/4” barbed fitting, 1/4” ID x 3/8” OD or 1/4” ID x 7/16” OD weldable C-Flex  tubing with a plugged end and tubing clamp.Supplied gamma irradiated, sterile and individually double bagged.