Learn More
Chemglass Life Sciences 5L Media Transfer Vessel, Vent Cap, w/Barb and Tubing, w/Quick Disconnect, Sterile
SDP
Catalog No. NC2038280
Encompass
Our 5L transfer flask is designed for either sterile transfer of liquid, or they are a great choice for a single use inoculation vessel.OUr media transfer vessel is ideal for use when doing a gravity feed from a sterile 5L vessel.Erlenmeyer Style Flasks with a 0. 2 um hydrophobic Filter, sterile and individually double bagged.The 5L Polycarbonate Flasks with Filter Cap has a 3/16" barbed fitting that accepts a 1/4" ID, 7/16" OD Platinum Cured Tubing with a Quick Disconnect.Supplied complete with a Tubing Clamp.Gamma irradiated, Sterile.
Catalog No. NC2038280 Supplier Chemglass Life Sciences Supplier No. CLS11715LQC
Our 5L transfer flask is designed for either sterile transfer of liquid, or they are a great choice for a single use inoculation vessel.OUr media transfer vessel is ideal for use when doing a gravity feed from a sterile 5L vessel.Erlenmeyer Style Flasks with a 0. 2 um hydrophobic Filter, sterile and individually double bagged.The 5L Polycarbonate Flasks with Filter Cap has a 3/16" barbed fitting that accepts a 1/4" ID, 7/16" OD Platinum Cured Tubing with a Quick Disconnect.Supplied complete with a Tubing Clamp.Gamma irradiated, Sterile.