Learn More
Ace Glass, Inc. 5000mL separatory funnel body only, pear shaped, #15Ace-Thred bottom, includes 6mm bore PTFE stopcock plug, 7223-42 #15 PTFE drip tip & 12632-38 size 38 PTFE stopper not included
SDP
Catalog No. 50974667
Encompass_Preferred
5000ml pear-shaped, separatory funnel body. Funnel has a 6mm bore PTFE stopcock plug and a #15 Ace-Thred bottom which creates a leak tight connection without the need for an o-ring.
Catalog No. 50-974-667 Supplier Ace Glass, Inc. Supplier No. 722337
5000ml pear-shaped, separatory funnel body. Funnel has a 6mm bore PTFE stopcock plug and a #15 Ace-Thred bottom which creates a leak tight connection without the need for an o-ring.