Learn More
Chemglass Life Sciences Flask, Round Bottom, 5000mL, Heavy Wall, 3-Neck, 45/50 CN - 29/42 SNs, Vertical, Jacketed
SDP
Catalog No. CG1538M36
Encompass_Preferred
Flask, Round Bottom, 5000mL, Heavy Wall, 45/50 - 29/42, 3-Neck, Vertical, Jacketed. Round bottom, HEAVY WALL, three-flask with standard taper outer joints on the center and side necks. Entire body of the flask is jacketed and has two hose connections for circulation of heating or cooling liquid. Indentations increase agitation of the contents of the flask during stirring.
Catalog No. CG1538M36 Supplier Chemglass Life Sciences Supplier No. CG1538M36
Flask, Round Bottom, 5000mL, Heavy Wall, 45/50 - 29/42, 3-Neck, Vertical, Jacketed. Round bottom, HEAVY WALL, three-flask with standard taper outer joints on the center and side necks. Entire body of the flask is jacketed and has two hose connections for circulation of heating or cooling liquid. Indentations increase agitation of the contents of the flask during stirring.