Learn More
Alkali Scientific 5L Roller Bottles 5000mL 1700c
SDP
Catalog No. 503026651
Encompass_Preferred
Our 5000mL Roller Bottle is designed for tissue culture applications It features printed graduations for easy volume measurement and comes with a vented cap to facilitate gas exchange Each bottle is sterile ensuring the integrity of your cell cultures With 12 bottles included in each case this product offers convenience and cost-effectiveness for your laboratory needs The roller bottles 170 cm size provides ample space for cell growth and harvesting Whether you are working on cell expansion virus propagation or protein production this roller bottle is a reliable choice for smooth and efficient tissue culture procedures Expanded Surface Roller Bottle increases surface area for cell culture Large knurled cap for easy grip Vented or non-vented cap for open or closed systems Manufactured from medical grade virgin polystyrene Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment Easy-to-read black printed graduations Packaged steri
Catalog No. 50-302-6651 Supplier Alkali Scientific Supplier No. VT5000P
Our 5000mL Roller Bottle is designed for tissue culture applications It features printed graduations for easy volume measurement and comes with a vented cap to facilitate gas exchange Each bottle is sterile ensuring the integrity of your cell cultures With 12 bottles included in each case this product offers convenience and cost-effectiveness for your laboratory needs The roller bottles 170 cm size provides ample space for cell growth and harvesting Whether you are working on cell expansion virus propagation or protein production this roller bottle is a reliable choice for smooth and efficient tissue culture procedures Expanded Surface Roller Bottle increases surface area for cell culture Large knurled cap for easy grip Vented or non-vented cap for open or closed systems Manufactured from medical grade virgin polystyrene Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment Easy-to-read black printed graduations Packaged steri